Cellulose chromatography papers
Sheets, 460×570 mm
Catalog # 552-0374
Supplier: Sartorius
Jump to:
Product Details & Documents
Chromatography papers are not only ideal for chromatography, but also for routine and repetitive separations, routine analysis of proteins in serum (e.g. human albumin) and several absorption applications like those common in the life sciences and diagnostics. They are made of cotton linters with more than 98% alpha-cellulose content and <0,04% ash content.
Specifications
- Description:Sheets
- Seq:16
- Size:460×570 mm
- Thickness:0,35 mm
- Cat. No.:552-0374
- Environmental certification:Renewable Energy Certificate
- Capillary rise:115 mm/30 min
- Grade:FN 100
- Environmentally Preferable:Low Manufacturing Impact
- Weight:195 g/m²



