Glass microfibre filters without binder
MGF, 90 mm
513-0078
Binder free glass microfibre filters are made of 100% borosilicate, are highly pure and extremely white. They are recommended for analytical and gravimetric analyses and also as prefilters. These filters combine fast flow rates with high load capacity and the retention of very fine particles.
- Biologically inert
- Chemical resistant
Grade MGA: Clarification of buffer and reagent solutions
Grade MG 160: Air monitoring
Grade MG 550-HA : Analysis of suspended solids in wastewater according to 2540D
Grade MGB, Grade MGD, Grade 13440: Membrane prefilter
Grade MGC: Analysis of suspended solids in waste according to DIN EN 872
Grade MGF: Clarification of protein solutions
Grade MGG: Water filtration
Documents
- Grade:MGF
- Seq:55
- Size:90 mm
- Thickness:0,40 mm
- Cat. No.:513-0078
- Filtration speed:185 s (Herzberg)
- Particle retention:0,7 µm
- Max. temperature:500 °C
- Weight:75 g/m²



