Microtitre plates, 96-well
V-Bottom, PS, Clear, 96, 280 µl
Catalog # 735-0184
Supplier: Thermo Fisher Scientific
Microtitre plates, 96-well
Catalog # 735-0184
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Rigid styrene or flexible vinyl, square well. Ideal for micro-volume applications, such as storage, sample or reagent transfer, serial dilution, haemogglutination, blood typing and compliment fixation. Available with a choice of flat, U- or V-bottom well shape.
- U-bottom well shape improves washing in ELISAs, enhances sensitivity in fluorescent applications and facilitates observation of agglutination reactions
- V-bottom well shape is optimal for magnetic bead assays, ideal for agglutination tests, useful for centrifugation or concentration and facilitates recovery of contents
Specifications
- No. of wells:96
- Colour:Clear
- Description:V-Bottom, PS
- Total volume:280 µl
- Cat. No.:735-0184
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact