0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
10 x REC Buffer 13 1ml 1 * 1 mL
10 x REC Buffer 13 1ml 1 * 1 mL
TREV3900-500-13
:
TREVIGEN
10 x REC Buffer 13 1ml 1 * 1 mL
TREV3900-500-13
:
TREVIGEN
10 x REC Buffer 13 1ml 1 * 1 mL
1 * 1 mL
-
+