Order Entry
Export
Orders LinkContactUsLinkComponent
Proteinase K

Proteinase K

Glass bottle, 1,25 mL
Catalog # AMBI2546
Supplier:   Ambion
Proteinase K
Catalog # AMBI2546
Supplier:  Ambion
CAS Number:  
Proteinase K

Glass bottle, 1,25 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1,25 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,25 mL
  • Cat. No.:
    AMBI2546
  • Enzyme activity:
    600 mAnsonU/ml
  • Form:
    Aqueous solution
  • Storage conditions:
    Refrigerator
  • Concentration:
    20 mg/ml
  • Enzyme name:
    Proteinase K
  • CAS No.:
    39450-01-6

Specifications