Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sample vials, E-C, WHEATON®

8,00 ml, 17×63 mm, Clear, Yes, Solid top, 15 - 425, Phenolic, Black, PTFE/14B rubber
  215-3578
Sample vials, E-C, WHEATON®
  215-3578
 :  
Sample vials, E-C, WHEATON®
Sample vials, E-C, WHEATON®

8,00 ml, 17×63 mm, Clear, Yes, Solid top, 15 - 425, Phenolic, Black, PTFE/14B rubber

 

Clear vials made from low extractable borosilicate glass that provides superior chemical resistance. Conform to USP Type I and ASTM E 438 Type I, Class A requirements.
Amber vials made from borosilicate glass that conforms to USP Type I requirements for light transmission to protect light-sensitive products.

 : Caps and replacement caps can be purchased separately.

 : Vials packed in two shrink-wrapped packs of 100 vials which reduces contamination from corrugate. The 60 ml vials are packed in one shrink-wrapped pack of 143 vials.

 
  • Cap included:
    Yes
  • Cap material:
    Phenolic
  • Cap Size:
    15 - 425
  • Ø×H:
    17×63 mm
  • Septum:
    PTFE/14B rubber
  • Capacity:
    8,00 ml
  • Colour:
    Clear
  • Seq:
    19
  • Cap type:
    Solid top
  • Cap colour:
    Black