Order Entry
Export
Orders LinkContactUsLinkComponent
 

Suction cannulae, Aesculap®

Suction cannula, Frazier, 2,3×110 mm
Catalog # AESCMD124R
Supplier:   Aesculap
Suction cannulae, Aesculap®
Catalog # AESCMD124R
Supplier:  Aesculap
Suction cannulae, Aesculap®

Suction cannula, Frazier, 2,3×110 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Suction cannulae with finger cut-off and stylet.

  • Author: Frazier
  • Thumb piece 90° to curve
  • With LUER hub
Specifications
  • Ø×L:
    2,3×110 mm
  • Description:
    Suction cannula, Frazier
  • Seq:
    01