Order Entry
Export
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Gouge for scoliose operating, Zielke, 5,5 mm, 230 mm
Catalog # AESCFL701R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL701R
Supplier:  Aesculap
Gouges, Aesculap®

Gouge for scoliose operating, Zielke, 5,5 mm, 230 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Type:
    Zielke
  • Width:
    5,5 mm
  • Description:
    Gouge for scoliose operating
  • Length:
    230 mm