Order Entry
Export
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Gouge for taking grafts fro the ribs, Walter, 15 mm, 205 mm
Catalog # AESCFL585R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL585R
Supplier:  Aesculap
Gouges, Aesculap®

Gouge for taking grafts fro the ribs, Walter, 15 mm, 205 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Type:
    Walter
  • Width:
    15 mm
  • Description:
    Gouge for taking grafts fro the ribs
  • Length:
    205 mm