Order Entry
Export
Orders LinkContactUsLinkComponent
 

Chisels, Aesculap®

Chisel, Partsch, 170 mm, 4 mm
Catalog # AESCFL184R
Supplier:   Aesculap
Chisels, Aesculap®
Catalog # AESCFL184R
Supplier:  Aesculap
Chisels, Aesculap®

Chisel, Partsch, 170 mm, 4 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

These chisels are used for dental surgery.

Specifications
  • Description:
    Chisel
  • Length:
    170 mm
  • Seq:
    10
  • Type:
    Partsch
  • Width:
    4 mm