Order Entry
Export
Orders LinkContactUsLinkComponent
Chisels, Aesculap®

Chisels, Aesculap®

Chisel, angled, -, 140 mm, 6 mm
Catalog # AESCFL122R
Supplier:   Aesculap
Chisels, Aesculap®
Catalog # AESCFL122R
Supplier:  Aesculap
Chisels, Aesculap®

Chisel, angled, -, 140 mm, 6 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

These chisels are used for dental surgery.

Specifications
  • Description:
    Chisel, angled
  • Length:
    140 mm
  • Seq:
    20
  • Type:
    -
  • Width:
    6 mm