Order Entry
Export
Orders LinkContactUsLinkComponent
Chisels, Aesculap®

Chisels, Aesculap®

Chisel, angled, -, 140 mm, 4 mm
  AESCFL120R
 :   Aesculap
Chisels, Aesculap®
  AESCFL120R
 :  Aesculap
Chisels, Aesculap®

Chisel, angled, -, 140 mm, 4 mm

 
 
   
1 * 1 items
 

These chisels are used for dental surgery.

 
  • Description:
    Chisel, angled
  • Length:
    140 mm
  • Seq:
    19
  • Type:
    -
  • Width:
    4 mm