Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bone curettes, Aesculap®

Bone curette, type Simon (Fig. 0), with 5,2 mm end, 235 mm
Catalog # AESCFK690R
Supplier:   Aesculap
Bone curettes, Aesculap®
Catalog # AESCFK690R
Supplier:  Aesculap
Bone curettes, Aesculap®

Bone curette, type Simon (Fig. 0), with 5,2 mm end, 235 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Bone curette, type Simon (Fig. 0), with 5,2 mm end
  • Length:
    235 mm
  • Seq:
    44
  • Cat. No.:
    AESCFK690R