Order Entry
Export
Orders LinkContactUsLinkComponent
Bone curettes, Aesculap®

Bone curettes, Aesculap®

Bone curette with PEEK-handle, Fig. 3, with 10 mm end, 270 mm
Catalog # AESCFK672R
Supplier:   Aesculap
Bone curettes, Aesculap®
Catalog # AESCFK672R
Supplier:  Aesculap
Bone curettes, Aesculap®

Bone curette with PEEK-handle, Fig. 3, with 10 mm end, 270 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Bone curette with PEEK-handle, Fig. 3, with 10 mm end
  • Length:
    270 mm
  • Seq:
    52