Order Entry
Export
Orders LinkContactUsLinkComponent
Bone curettes, Aesculap®

Bone curettes, Aesculap®

Bone curette, Willinger type (Fig. 00), with 3,4 mm end, 170 mm
Catalog # AESCFK658R
Supplier:   Aesculap
Bone curettes, Aesculap®
Catalog # AESCFK658R
Supplier:  Aesculap
Bone curettes, Aesculap®

Bone curette, Willinger type (Fig. 00), with 3,4 mm end, 170 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Bone curette, Willinger type (Fig. 00), with 3,4 mm end
  • Length:
    170 mm
  • Seq:
    25