Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bone curettes, Aesculap®

Bone curette, Volkmann type (Fig. 000), with 3,6 mm end, 170 mm
  AESCFK631R
 :   Aesculap
Bone curettes, Aesculap®
  AESCFK631R
 :  Aesculap
Bone curettes, Aesculap®

Bone curette, Volkmann type (Fig. 000), with 3,6 mm end, 170 mm

 
 
   
1 * 1 items
 
 
  • Description:
    Bone curette, Volkmann type (Fig. 000), with 3,6 mm end
  • Length:
    170 mm
  • Seq:
    16