Micro scissors for endoscopic work, Aesculap® MINOP®
Replacement inner tube for MINOP instrument FF385R
AESCFF435R
: Aesculap
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
- Very delicate
Ø = 2 mm
: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
- Length:Replacement inner tube for MINOP instrument FF385R