Orders LinkContactUsLinkComponent
Micro scissors for endoscopic work, Aesculap® MINOP®
Micro scissors for endoscopic work, Aesculap® MINOP®
Replacement outer tube for MINOP instruments FF385R - FF389R
Catalog # AESCFF433R
Supplier:   Aesculap
Micro scissors for endoscopic work, Aesculap® MINOP®
Catalog # AESCFF433R
Supplier:  Aesculap
Micro scissors for endoscopic work, Aesculap® MINOP®
Replacement outer tube for MINOP instruments FF385R - FF389R
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
  • Very delicate
Ø = 2 mm
Ordering information: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
Specifications
  • Length:
    Replacement outer tube for MINOP instruments FF385R - FF389R