Orders LinkContactUsLinkComponent
Micro scissors for endoscopic work, Aesculap® MINOP®
Micro scissors for endoscopic work, Aesculap® MINOP®
FF385R, sharp/sharp, 265 mm
Catalog # AESCFF385R
Supplier:   Aesculap
Micro scissors for endoscopic work, Aesculap® MINOP®
Catalog # AESCFF385R
Supplier:  Aesculap
Micro scissors for endoscopic work, Aesculap® MINOP®
FF385R, sharp/sharp, 265 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
  • Very delicate
Ø = 2 mm
Ordering information: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
Specifications
  • Type:
    FF385R, sharp/sharp
  • Length:
    265 mm
  • Seq:
    02