Order Entry
Export
Orders LinkContactUsLinkComponent
Micro suture tying forceps, Aesculap®

Micro suture tying forceps, Aesculap®

Straight, 0,5 mm jaws, 180 mm
Catalog # AESCFD286R
Supplier:   Aesculap
Micro suture tying forceps, Aesculap®
Catalog # AESCFD286R
Supplier:  Aesculap
Micro suture tying forceps, Aesculap®

Straight, 0,5 mm jaws, 180 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

These micro suture tying forceps feature a round handle.

Specifications
  • Type:
    Straight, 0,5 mm jaws
  • Length:
    180 mm
  • Seq:
    05