Micro suture tying forceps, Aesculap®
Curved, 0,3 mm jaws, 120 mm
AESCFD281R
: Aesculap
These micro suture tying forceps feature a round handle.
- Type:Curved, 0,3 mm jaws
- Length:120 mm
- Seq:02
These micro suture tying forceps feature a round handle.