Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Forceps
Dissecting forceps, S-shaped, Aesculap®
Dissecting forceps, S-shaped, Aesculap®
Print
Dissecting forceps, S-shaped, Aesculap®
S-shaped, delicate, 210 mm
Catalog # AESCBJ016R
Supplier:
Aesculap
Dissecting forceps, S-shaped, Aesculap®
Catalog # AESCBJ016R
Supplier:
Aesculap
Expand image
Return to Top
Dissecting forceps, S-shaped, Aesculap®
S-shaped, delicate, 210 mm
Specifications
Add to cart
Jump to:
Specifications
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Type:
S-shaped, delicate
Length:
210 mm
Seq:
02