Orders LinkContactUsLinkComponent
Surgical scissors, Aesculap®
Surgical scissors, Aesculap®
Straight, sharp/sharp, 130 mm
Catalog # AESCBC343R
Supplier:   Aesculap
Surgical scissors, Aesculap®
Catalog # AESCBC343R
Supplier:  Aesculap
Surgical scissors, Aesculap®
Straight, sharp/sharp, 130 mm
Price
35,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These straight scissors are ideal for dental surgery.
Specifications
  • Type:
    Straight, sharp/sharp
  • Length:
    130 mm
  • Seq:
    05