Orders LinkContactUsLinkComponent
Pots, laboratory, without lid
Pots, laboratory, without lid
Lids for laboratory pots, stainless steel 18/10, for 5 l pots
Catalog # BOCH8372
Supplier:   BOCHEM
Pots, laboratory, without lid
Catalog # BOCH8372
Supplier:  BOCHEM
Pots, laboratory, without lid
Pots, laboratory, without lid
Lids for laboratory pots, stainless steel 18/10, for 5 l pots
Price
32,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel 18/10.
  • Rounded corners for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
Multipurpose pots for a wide range of uses. This series is suitable for induction fields.
Ordering information: Lids to be ordered separately.
Specifications
  • Description:
    Lids for laboratory pots, stainless steel 18/10, for 5 l pots
  • Seq:
    11