Order Entry
Export
Orders LinkContactUsLinkComponent
Measuring beakers, conical, graduated

Measuring beakers, conical, graduated

500 ml, 95 mm, 125 mm
Catalog # BOCH8650
Supplier:   BOCHEM
Measuring beakers, conical, graduated
Catalog # BOCH8650
Supplier:  BOCHEM
Measuring beakers, conical, graduated
Measuring beakers, conical, graduated

500 ml, 95 mm, 125 mm

Price
23,60 €
Log in for availability
1 * 1 items
Product Details & Documents

Stainless steel, 18/10, with handle and spout.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Ext.Ø:
    125 mm
  • Capacity:
    500 ml
  • Height:
    95 mm