Order Entry
Export
Orders LinkContactUsLinkComponent
 

Double bossheads

Double bosshead, with socket screw M6/M8, 16,5 mm
Catalog # BOCH5310
Supplier:   BOCHEM
Double bossheads
Catalog # BOCH5310
Supplier:  BOCHEM
Double bossheads
Double bossheads

Double bosshead, with socket screw M6/M8, 16,5 mm

Price
53,60 €
Log in for availability
1 * 1 items
Product Details & Documents

Double bossheads made of stainless steel 18/10.

  • For rods up to 16,5 mm
  • Angle: 90°
  • Available with heart screw M6/M8, socket screw M6/M8 or safety screw type 'heart' M8

 : DIN 12895

Specifications
  • Description:
    Double bosshead, with socket screw M6/M8
  • For rod Ø:
    16,5 mm