Order Entry
Export
Orders LinkContactUsLinkComponent
 

Double bossheads, cross type

Cast iron, 16,5 mm
  BOCH5366
 :   BOCHEM
Double bossheads, cross type
  BOCH5366
 :  BOCHEM
Double bossheads, cross type
Double bossheads, cross type

Cast iron, 16,5 mm

 
17,00 €
   
1 * 1 items
 

Cross type double bossheads, either made of stainless steel or malleable cast iron, chromium plated.

  • For rods up to 16,5 mm
  • Angle: 90 + 180°
  • With safety screw type 'heart' (M6)
 
  • Material:
    Cast iron
  • For rod Ø:
    16,5 mm