Order Entry
Export
Orders LinkContactUsLinkComponent
Bossheads with hook

Bossheads with hook

Steel, nickel plated, 13 mm
Catalog # BOCH5364
Supplier:   BOCHEM
Bossheads with hook
Catalog # BOCH5364
Supplier:  BOCHEM
Bossheads with hook
Bossheads with hook

Steel, nickel plated, 13 mm

Price
11,40 €
Log in for availability
1 * 1 items
Product Details & Documents

Bossheads with hook, either made of stainless steel or nickel plated steel.

  • For rods up to 13 mm
  • Angle: 90°
Specifications
  • Material:
    Steel, nickel plated
  • For rod Ø:
    13 mm