Mounting/reducer clips for tubes
Mounting clip/reducer clamp (OD 26,9 mm, Ø 12 mm), Tubes with different diameters
BOCH554
: BOCHEM
Mounting clips/reducers made of stainless steel. Either for tubes with identical diameter or for tubes with different diameters.
- For:Tubes with different diameters
- Description:Mounting clip/reducer clamp (OD 26,9 mm, Ø 12 mm)