Orders LinkContactUsLinkComponent
Instrument trays
Instrument trays
288×428×17 mm
Catalog # BURK4230-3043
Supplier:   BURKLE
Instrument trays
Catalog # BURK4230-3043
Supplier:  BURKLE
Instrument trays
Instrument trays
288×428×17 mm
Price
35,70 €
Log in for availability
1 * 1 items
Product Details & Documents
Melamine, white.
  • Tray without lid
    Suitable for use with foodstuffs
    Not autoclavable
    Not suitable for high temperatures
Ordering information: Lid available as accessory
Specifications
  • W×D×H:
    288×428×17 mm