Instrument trays
208×268×17 mm
Catalog # BURK4230-3027
Supplier: BURKLE
Jump to:
Product Details & Documents
Melamine, white.
Tray without lid
Suitable for use with foodstuffs
Not autoclavable
Not suitable for high temperatures
Ordering information: Lid available as accessory
Specifications
- W×D×H:208×268×17 mm