Order Entry
Export
Orders LinkContactUsLinkComponent
Pipette tips, Finntip™

Pipette tips, Finntip™

100 - 1000 µl, -, 71 mm, Blue, 10×96/refill
Catalog # 613-4485
   Low Manufacturing Impact
Pipette tips, Finntip™
Catalog # 613-4485
   Low Manufacturing Impact
Pipette tips, Finntip™
Pipette tips, Finntip™

100 - 1000 µl, -, 71 mm, Blue, 10×96/refill

Price
81,10 €
Log in for availability
1 * 960 items
Product Details & Documents

Finntip™ pipette tips are designed to achieve the highest level of performance in terms of accuracy and repeatability in conjunction with Finnpipette™ pipettes.

  • High quality PP, autoclavable
  • Unique shape of the collar ensures a leak-free connection to the pipette
  • Smooth, hydrophobic surface minimises liquid retention allowing total dispensing of the sample
  • Colour coded racks for rapid identification of the tips to be used
  • Sterile tips: Certified RNase-, DNase-, DNA- and endotoxin-free

Packaging: Available in different volumes and packed loose, in racks, in a starter kit (racks and refills) or as a refill kit.

Specifications
  • Colour code:
    Blue
  • Packed:
    10×96/refill
  • Sterile:
    -
  • Volume:
    100 - 1000 µl
  • Length:
    71 mm
  • Seq:
    29
  • Cat. No.:
    613-4485
  • Environmental certification:
    ISO 14001 (Certificate Registration No. EMS 623400)
  • Environmentally Preferable:
    Low Manufacturing Impact