Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TEMPERATURE PROBE GTF1200 -200/1150°C 1 * 1 items
Catalog # GRSG600507
Supplier:
Greisinger
Expand image
TEMPERATURE PROBE GTF1200 -200/1150°C 1 * 1 items
Catalog # GRSG600507
Supplier:
Greisinger