Order Entry
Export
Orders LinkContactUsLinkComponent
Toluene ≥98%, TECHNICAL

Toluene ≥98%, TECHNICAL

Plastic bottle, fluorinated, 5 L
Catalog # 28701.364
Flammable
Health
Irritant
Supplier:   VWR Chemicals
Toluene ≥98%, TECHNICAL
Catalog # 28701.364
Supplier:  VWR Chemicals
CAS Number:  108-88-3
Toluene ≥98%, TECHNICAL
Toluene ≥98%, TECHNICAL

Plastic bottle, fluorinated, 5 L

CAS Number:  108-88-3
Price
118,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Methylbenzene, Phenylmethane, FM approval
  • CAS Number:108-88-3
  • Formula:C₆H₅CH₃
  • MDL Number:MFCD00008512
  • Molecular Weight:92.14 g/mol
  • Merck Index:13,09607
  • Boiling Pt:110.6 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:0.867 g/cm³ (20 °C)
  • Flash Pt:4 °C
  • Storage Temperature:Ambient
  • EINECS:203-625-9
  • UN:1294
  • ADR:3,II
  • REACH:01-2119471310-51
Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specifications

  • Pack type:
    Plastic bottle, fluorinated
  • Pk:
    5 L

Specifications