Methanol ≥98.5%, TECHNICAL
Plastic drum, 25 L
Supplier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Catalog # 20903.461
Supplier: VWR Chemicals
CAS Number: 67-56-1
Jump to:
Product Details & Documents
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64.6 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.7918 g/cm³ (20 °C)
- Flash Pt:11 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3 (6.1),II
- REACH:01-2119433307-44
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951065)
Specifications
Specification Test Results
- Assay:Min. 98.5 %
- Evaporation residue:Max. 0.015 %
Specifications
- Pack type:Plastic drum
- Pk:25 L
- Cat. No.:20903.461
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



