Order Entry
Export
Orders LinkContactUsLinkComponent
Casting dams, Owl™

Casting dams, Owl™

Casting dam, 13 mm (L), DAM-13
Catalog # OWLADAM-13
Casting dams, Owl™
Catalog # OWLADAM-13
Casting dams, Owl™
Casting dams, Owl™

Casting dam, 13 mm (L), DAM-13

Price
184,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Model:
    DAM-13
  • Description:
    Casting dam, 13 mm (L)
  • Seq:
    01
  • Cat. No.:
    OWLADAM-13
  • Gel size W×L:
    130×130 mm