Robotic tips for Beckman®
919-021-05 (Low Retention), 130 µl, +, +, Transparent, 10 racks, 96 each
Catalog # 736-0093
Supplier: Thermo Fisher Scientific
Robotic tips for Beckman®
Catalog # 736-0093
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
High quality tips, tested on the workstations for which they were created, to ensure compatibility and a secure fit. Certified free of RNase, DNase, DNA, RNA, pyrogens and ATP.
- Transparent PP
- Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
- Low retention tips: Hydrophobic tips retain less sample which decreases the amount of reagent loss while significantly increasing result reproducibility
- Sterile tips: Sterilised by gamma radiation
: * Requires adapter 3772 only available directly from Molecular BioProducts.
Certificates
Search by Lot NumberSpecifications
- Packed:10 racks, 96 each
- Sterile:+
- Volume:130 µl
- Tip style:919-021-05 (Low Retention)
- Tip colour:Transparent
- Filter:+
- Cat. No.:736-0093
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact



