Pipette tips, ART™
ART™ 20 SoftFit® Pure Low Retention, 20 µl, +, 10 racks, 96 each
Catalog # 732-0327
Supplier: Thermo Fisher Scientific
Pipette tips, ART™
Catalog # 732-0327
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Transparent PP, sterile or non sterile.
- Available as non sterile bulk or racked; and as sterile racked
- Certified sterile tips, free of DNase, RNase, DNA and pyrogens (sterilised by gamma radiation)
- SoftFit® design assures a consistent, reliable fit for all channels of the pipette, which in turn ensures reliable distribution of the liquid
: Supplied loose bagged or in racks.
Specifications
- Packed:10 racks, 96 each
- Sterile:+
- Volume:20 µl
- Seq:15
- Tip style:ART™ 20 SoftFit® Pure Low Retention
- Cat. No.:732-0327
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact