Orders LinkContactUsLinkComponent
Hydrophobic interaction chromatography columns, TSK-GEL®
Hydrophobic interaction chromatography columns, TSK-GEL®
Guardgel kit, for P/Ns 14013 and 14014, Guard column, Ether-5PW, 20 µm
Catalog # TOYO0014025
Supplier:   TOSOH Bioscience
Hydrophobic interaction chromatography columns, TSK-GEL®
Catalog # TOYO0014025
Supplier:  TOSOH Bioscience
Hydrophobic interaction chromatography columns, TSK-GEL®
Hydrophobic interaction chromatography columns, TSK-GEL®
Guardgel kit, for P/Ns 14013 and 14014, Guard column, Ether-5PW, 20 µm
Price
1 160,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
These are polymethacrylate-based columns with a choice of three ligands (ether, phenyl or butyl) with varied hydrophobicities from low to high. Suitable for LC-MS applications.
  • Rigid polymeric base resin
  • Eliminates undesirable interactions with column hardware
  • Wide pH range (2 to 12) enables robust cleaning options
Documents
Specifications
  • Column Format:
    Guard column
  • Particle Size:
    20 µm
  • Description:
    Guardgel kit, for P/Ns 14013 and 14014
  • Phase:
    Ether-5PW