Hydrophobic interaction chromatography columns, TSK-GEL®
Guardgel kit, for P/N 07573, Guard column, Phenyl-5PW, 20 µm
Catalog # TOYO0007652
Supplier: TOSOH Bioscience
Hydrophobic interaction chromatography columns, TSK-GEL®
Catalog # TOYO0007652
Supplier: TOSOH Bioscience
Jump to:
Product Details & Documents
These are polymethacrylate-based columns with a choice of three ligands (ether, phenyl or butyl) with varied hydrophobicities from low to high. Suitable for LC-MS applications.
- Rigid polymeric base resin
- Eliminates undesirable interactions with column hardware
- Wide pH range (2 to 12) enables robust cleaning options
Certificates
Search by Lot NumberSpecifications
- Column Format:Guard column
- Particle Size:20 µm
- Description:Guardgel kit, for P/N 07573
- Phase:Phenyl-5PW