Ethanol ≥96% (v/v), TechniSolv®, pure
Plastic drum, 10 L
Ethanol ≥96% (v/v), TechniSolv®, pure
83804.410
: 64-17-5
: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
: Ethyl alcohol
- :64-17-5
- :C₂H₅OH
- :MFCD00003568
- :46.07 g/mol
- :78.3 °C (1013 hPa)
- :–117 °C
- :0.7895 g/cm³ (20 °C)
- :12 °C (closed cup)
- :Ambient
- :200-578-6
- :1170
- :3,II
- :01-2119457610-43
10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
- Assay:Min. 95 %
- Assay (V/V):Min. 95 %
- Density (20/20):0.805 - 0.811
- Pack type:Plastic drum
- Pk:10 L
- Cat. No.:83804.410
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Safer Human and Environmental Health



