Order Entry
Export
Orders LinkContactUsLinkComponent
Methanol

Methanol

Methanol
Catalog # EHERC14995000
Supplier:   EHRENSTORFER
Methanol
Catalog # EHERC14995000
Supplier:  EHRENSTORFER
CAS Number:  
Methanol
Methanol

Methanol

CAS Number:  
Price
21,40 €
Log in for availability
1 * 1 mL
Product Details & Documents
  • SMILES: CO
  • Inchi: InChI=1S/CH4O/c1-2/h2H,1H3
  • IUPAC: methanol
  • Packing group: II

Documents

Specifications

Specifications

  • Description:
    Methanol
  • UN No.:
    1230
  • Cat. No.:
    EHERC14995000
  • Formula:
    CH₄O
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS02, GHS06, GHS08
  • GHS signal:
    Danger
  • Synonym(s):
    Methanol;Methyl alcohol
  • Molecular weight:
    32,04
  • CAS No.:
    67-56-1
  • Shipping conditions:
    RT

Specifications