Atrazine
Atrazine
Catalog # EHERC10330000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCNc1nc(Cl)nc(NC(C)C)n1
- Inchi: InChI=1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)
- IUPAC: 6-chloro-4-N-ethyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
- Packing group: III
Specifications
Specifications
- Description:Atrazine
- UN No.:3077
- Cat. No.:EHERC10330000
- Formula:C₈H₁₄ClN₅
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Warning
- Synonym(s):1,3,5-Triazine-2,4-diamine, 6-chloro-N2-ethyl-N4-(1-methylethyl)-;1,3,5-Triazine-2,4-diamine, 6-chloro-N-ethyl-N'-(1-methylethyl)- (9CI);s-Triazine, 2-chloro-4-(ethylamino)-6-(isopropylamino)- (8CI);6-Chloro-N2-ethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine;1-Chloro-3-ethylamino-5-isopropylamino-2,4,6-triazine;2-Chloro-4-(ethylamino)-6-(2-propylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)triazine;2-Chloro-4-ethylamineisopropylamine-s-triazine;2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-s-triazine;6-Chloro-4-(ethylamino)-2-(isopropylamino)-s-triazine;6-Chloro-N-Ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine;A 361;AAtrex;AAtrex 4L;AAtrex Nine-O;ATZ;Aatrax;Aatrex 4LC;Aatrex L;Akticon;Aktikon;Aktikon PK;Aktinit A;Aktinit PK;Argezin;Atradex;Atragranz;Atranex;Atranex 80WP;Atraphyt;Atrasan;Atrataf;Atrazerba FL;Atrazin;Atrazine;Azoprim;Azoprim 50;CET;Chromozin;Cyazin;Farmozine;G 30027;Gesamprim;Gesaprim;Gesaprim 50;Gesaprim 500;Gesaprim L;Herbatoxol;Hungazin;Hungazin PK;NSC 163046;Nortox 500SC;Nu-Trazine;Oleogesaprim;Oleogesaprim 200;Primatol A;Primoleo;Radazin;Radazin T;Triazine A 1294;Wonuk;Zealin L;Zeapos;Zeazin;Zeazine;Zeopos
- Molecular weight:215,68
- CAS No.:1912-24-9
- Shipping conditions:RT



