Orders LinkContactUsLinkComponent
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
Iodine colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, iodine discs, W680/OG/25 and W680/OG/10 mm fused cells, AF333
Catalog # 633-0153
Supplier:   Lovibond Tintometer
Comparator kits, Lovibond® system 2000
Catalog # 633-0153
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
Iodine colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, iodine discs, W680/OG/25 and W680/OG/10 mm fused cells, AF333
Price
3 070,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Available for many of the commonly used colour scales, these kits are a convenient means of ordering the complete range of equipment required. Each system includes.
  • A visual comparator instrument which brings the sample and colour standards into the same field of view for more accurate colour matching
  • A selection of colour standards - stable-coloured glass filters in test discs, which have been pre-calibrated to give direct readings over set measuring ranges
  • A standardised benchtop light source to ensure constant, uniform lighting conditions for colour matching and, therefore, consistent and accurate readings
  • Glassware of the appropriate type and path length
Specifications
  • Description:
    Iodine colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, iodine discs, W680/OG/25 and W680/OG/10 mm fused cells
  • Type:
    AF333