Orders LinkContactUsLinkComponent
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
IFU colour scale colour grading kit for fruit juices, including Colour Comparator 2000+ with Daylight 2000 Lighting unit,FU discs 4/48,1 - 9; 4/49 9 - 17; 4/50 17 - 25), W680/OG/13 mm fused cells, AF331
Catalog # 633-0152
Supplier:   Lovibond Tintometer
Comparator kits, Lovibond® system 2000
Catalog # 633-0152
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
IFU colour scale colour grading kit for fruit juices, including Colour Comparator 2000+ with Daylight 2000 Lighting unit,FU discs 4/48,1 - 9; 4/49 9 - 17; 4/50 17 - 25), W680/OG/13 mm fused cells, AF331
Price
2 930,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Available for many of the commonly used colour scales, these kits are a convenient means of ordering the complete range of equipment required. Each system includes.
  • A visual comparator instrument which brings the sample and colour standards into the same field of view for more accurate colour matching
  • A selection of colour standards - stable-coloured glass filters in test discs, which have been pre-calibrated to give direct readings over set measuring ranges
  • A standardised benchtop light source to ensure constant, uniform lighting conditions for colour matching and, therefore, consistent and accurate readings
  • Glassware of the appropriate type and path length
Specifications
  • Description:
    IFU colour scale colour grading kit for fruit juices, including Colour Comparator 2000+ with Daylight 2000 Lighting unit,FU discs 4/48,1 - 9; 4/49 9 - 17; 4/50 17 - 25), W680/OG/13 mm fused cells
  • Type:
    AF331