Order Entry
Export
Orders LinkContactUsLinkComponent
Pressure vessels

Pressure vessels

4,54 l, Dispensing pressure vessel, for holding liquid for filtration
Catalog # MILFXX6700P01
Supplier:   Merck
Pressure vessels
Catalog # MILFXX6700P01
Supplier:  Merck
Pressure vessels
Pressure vessels

4,54 l, Dispensing pressure vessel, for holding liquid for filtration

Price
1 260,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Dispensing pressure vessels hold liquid for filtration through pressure-operated filter holders, if connected to an external pressure source. Used for large-volume filtration, reservoir for buffer or solvent dispensing.

  • Made from stainless steel 316
  • Closures are secured by cam-lock handle
  • Vessels are autoclavable when vented
  • Fluoroelastomer gaskets and O-rings used
  • Molded neoprene rubber base

Max. operating pressure: 6,9 bar

Max. operating temperature: 121 °C

Certifications: Meets ASME®-UM Code requirements.

Specifications
  • Capacity:
    4,54 l
  • Description:
    Dispensing pressure vessel, for holding liquid for filtration
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • O-ring, fluoroelastomer A
    O-ring, fluoroelastomer A
    MILFXX6700059
    Log in for availability
    1 * 1 items - 155,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Bubble point test apparatus
    Bubble point test apparatus
    MILFXX6700L55
    Log in for availability
    1 * 1 items - 1 560,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Pressure tubing, PVC, I.Ø 12,7 mm
    Pressure tubing, PVC, I.Ø 12,7 mm
    MILFXX6700035
    Log in for availability
    1 * 1 items - 131,75 €

Product Information

  • Catalog #
    Availability
    Price
  • Pressure vessel accessory kit
    Pressure vessel accessory kit
    MILFXX67000PK
    Log in for availability
    1 * 1 items - 1 960,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Plug, stainless steel, 6,35 mm NPT
    Plug, stainless steel, 6,35 mm NPT
    MILFYY1301009
    Log in for availability
    1 * 1 items - 60,10 €

Product Information

  • Catalog #
    Availability
    Price
  • Quick-release nipple and coupling, 6,35 mm NPTM
    Quick-release nipple and coupling, 6,35 mm NPTM
    MILFXX6700030
    Log in for availability
    1 * 1 items - 200,60 €

Product Information

  • Catalog #
    Availability
    Price
  • Pressure gauge, 6,35 mm ASME
    Pressure gauge, 6,35 mm ASME
    MILFXX6700L15
    Log in for availability
    1 * 1 items - 260,95 €

Product Information

  • Catalog #
    Availability
    Price
  • Vent/relief valve, 7 bar, ASME
    Vent/relief valve, 7 bar, ASME
    MILFXX6700L24
    Log in for availability
    1 * 1 items - 291,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter, M, 6,35 mm NPT to 6,35 - 12,7 mm I.Ø
    Adapter, M, 6,35 mm NPT to 6,35 - 12,7 mm I.Ø
    MILFXX6700L11
    Log in for availability
    1 * 2 items - 262,00 €