Orders LinkContactUsLinkComponent
Caps, Versilic®
Caps, Versilic®
12,0 mm
Catalog # 217-0002
Caps, Versilic®
Catalog # 217-0002
Caps, Versilic®
Caps, Versilic®
12,0 mm
Price
176,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Made of translucent silicone. Ideal for protecting piping/tubing or protruding cylindrical components during transport and all operations that require aperture protection. (against knocks, splashes, paint and more).
  • Outstanding sealing and protection from the outside
  • Can be perforated and autoclaved
  • Resistant to ageing, ozone and repeated sterilisation
The size given corresponds to the Ext. Ø of the vessel to be capped in mm. Resistant to temperatures from −60 to +220 °C (up to 250 °C for short periods).
Documents
Specifications
  • For Ø ext.:
    12,0 mm
  • Seq:
    02