Microplates, 1536-well
Solid bottom, black, FLUOTRAC 200, Black, 1536, -
Catalog # 738-0084
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or white in LUMITRAC™ or FLUOTRAC™ quality.
- HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
- Solid bottom plates available with high binding (MICROLON™ 600, LUMITRAC™ 600 or FLUOTRAC™ 600) or medium binding (LUMITRAC™ 200 or FLUOTRAC™ 200) surfaces
- Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl) with the advantages of a round well (reduced wicking and bubbling)
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Working volume: 3 - 10 µl
Documents
Specifications
- No. of wells:1536
- Sterile:-
- Colour:Black
- Description:Solid bottom, black, FLUOTRAC 200
- Cat. No.:738-0084
- Environmental certification:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact