Polishing cloths, KIMTECH PREP*
White, 600×390 mm, Refill for bucket, 1 roll of 300 sheets
Catalog # 115-2199
Supplier: KIMBERLY CLARK
Jump to:
Product Details & Documents
The KIMTECH PREP* polishing wipes are ideal spunlace products for dry polishing glass and other materials. They are also suitable for pre- and post-paint applications.
- Large centrefeed rolls in dispensing bucket
- Soft and bulky - no scratching
Specifications
- Packed:Refill for bucket, 1 roll of 300 sheets
- Colour:White
- L×W:600×390 mm