Storage plates, 384-well, MASTERBLOCK®
PP, V-bottom, non-sterile, without lid, deep well MASTERBLOCK®, Natural
Catalog # 736-0151
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
PP, without lid. Ideal for compound libraries and sample storage.
- Biologically inert, resistant to commonly used solvents (such as DMSO, acetonitrile or acetone) and temperature resistant (−196 °C to +121 °C)
- Uniform external dimensions and tight tolerances make robotic handling easier
- Can be easily sealed with adhesive films, heat seals or CapMats
- Alphanumeric well coding
- Conical well shape enables precise pipetting and rounded square well design combines flexible working volume (20 to 225 µl) with reduced wicking and bubbling
Colour: Natural
Colour: Natural
Documents
Specifications
- Colour:Natural
- Description:PP, V-bottom, non-sterile, without lid, deep well MASTERBLOCK®



